missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Rhenium Standard for ICP 1000 ppm in H2O, Ricca Chemical

Catalog No. p-7074095
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010306100 100 mL
RV010306250 250 mL
RV010306500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010306100 Supplier Ricca Chemical Company Supplier No. RV010306100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 13598-65-7 , 7732-18-5
Molecular Formula H4NO4Re
Molecular Weight (g/mol) 268.24
MDL Number MFCD00012377
InChI Key XPSXHXWHJAIQJR-UHFFFAOYSA-M
PubChem CID 3084163
IUPAC Name rheniumoylol amine
SMILES N.O[Re](=O)(=O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.86%, 0.14%
CAS Max % 100%, 0.14%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity