missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Rhenium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$1360.37 - $4166.60
Chemical Identifiers
| CAS | 13768-11-1 |
|---|---|
| Molecular Formula | HO4Re |
| Molecular Weight (g/mol) | 251.21 |
| MDL Number | MFCD00011326 |
| InChI Key | UGSFIVDHFJJCBJ-UHFFFAOYSA-M |
| PubChem CID | 83718 |
| IUPAC Name | rheniumoylol |
| SMILES | O[Re](=O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010350100
|
Ricca Chemical Company
RV010350100N |
100 mL |
Each for $1,360.37
|
|
|||||
|
RV010350250
|
Ricca Chemical Company
RV010350250N |
250 mL |
Each for $2,553.67
|
|
|||||
|
RV010350500
|
Ricca Chemical Company
RV010350500N |
500 mL |
Each for $4,166.60
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 13768-11-1 | |
| 251.21 | |
| UGSFIVDHFJJCBJ-UHFFFAOYSA-M | |
| rheniumoylol |
| HO4Re | |
| MFCD00011326 | |
| 83718 | |
| O[Re](=O)(=O)=O |
Specifications
| Natural LDPE Bottle | |
| 95.91%,1.96%,0.13% | |
| 1 | |
| HO4Re | |
| Approximately 0°C | |
| UGSFIVDHFJJCBJ-UHFFFAOYSA-M | |
| rheniumoylol | |
| 100 mL | |
| High Purity |
| 13768-11-1 , 7697-37-2 , 7732-18-5 | |
| 99.83%,2.04%,0.13% | |
| Liquid | |
| MFCD00011326 | |
| Standard | |
| O[Re](=O)(=O)=O | |
| 251.21 | |
| 83718 |