missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Rhenium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010350100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010350100 100 mL
RV010350250 250 mL
RV010350500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010350100 Supplier Ricca Chemical Company Supplier No. RV010350100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 13768-11-1 , 7697-37-2 , 7732-18-5
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
PubChem CID 83718
IUPAC Name rheniumoylol
SMILES O[Re](=O)(=O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.91%, 1.96%, 0.13%
CAS Max % 99.83%, 2.04%, 0.13%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity