missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ QC Standard, Ricca Chemical

Catalog No. RV010694500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010694500 500 mL
RV010694100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010694500 Supplier Ricca Chemical Company Supplier No. RV010694500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for Chlorines/Chlorides 10,000 mg/L /nManufactured and Tested in an ISO 17025/Guide 34 Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
CAS Min % 1.62%, 0.98%
CAS Max % 1.68%, 1.02%
Specific Gravity 1.06
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity