missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Praseodymium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$467.57 - $1,192.10
Chemical Identifiers
| CAS | 15878-77-0 |
|---|---|
| Molecular Formula | H12N3O15Pr |
| Molecular Weight (g/mol) | 435.01 |
| InChI Key | LXXCECZPOWZKLC-UHFFFAOYSA-N |
| PubChem CID | 204170 |
| IUPAC Name | praseodymium(3+) hexahydrate trinitrate |
| SMILES | O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010303100
|
Ricca Chemical Company
RV010303100N |
100 mL |
N/A
|
|
|||||
|
RV010303250
|
Ricca Chemical Company
RV010303250N |
250 mL |
N/A
|
|
|||||
|
RV010303500
|
Ricca Chemical Company
RV010303500N |
500 mL |
N/A
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 15878-77-0 | |
| 435.01 | |
| 204170 | |
| O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| H12N3O15Pr | |
| LXXCECZPOWZKLC-UHFFFAOYSA-N | |
| praseodymium(3+) hexahydrate trinitrate |
Specifications
| Natural LDPE Bottle | |
| 95.74%,1.96%,0.30% | |
| 1 | |
| H12N3O15Pr | |
| Standard | |
| O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 435.01 | |
| 204170 |
| 15878-77-0 , 7697-37-2 , 7732-18-5 | |
| 99.64%,2.04%,0.32% | |
| Liquid | |
| Approximately 0°C | |
| LXXCECZPOWZKLC-UHFFFAOYSA-N | |
| praseodymium(3+) hexahydrate trinitrate | |
| 100 mL | |
| High Purity |