missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Palladium Standard for ICP 10 g/L 5% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010474100N

Numéro de catalogue. RV010474100


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • 10 g/L 5% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Spécifications

Spécifications

Palladium ICP Standard
10102-05-3,7697-37-2,7732-18-5
94.70%,5.10%,2.21%
Liquid
MFCD00011169
Colorless
Approximately 0°C
Miscible with water
Standard
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
230.43
24932
Natural LDPE Bottle
90.98%,4.90%,2.13%
1
N2O6Pd
UN3264
<2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
GPNDARIEYHPYAY-UHFFFAOYSA-N
palladium(2+) dinitrate
100 mL
High Purity
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions