missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Palladium Standard for ICP 1000 ppm in 5% HCl, Ricca Chemical

Catalog No. rv010302500
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010302500 500 mL
RV010302250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010302500 Supplier Ricca Chemical Company Supplier No. RV010302500N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 5% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10102-05-3 , 7647-01-0 , 7732-18-5
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
PubChem CID 24932
IUPAC Name palladium(2+) dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 92.93%, 4.90%
CAS Max % 96.73%, 5.10%
Specific Gravity 1
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity