missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Nitrite(N) - NO2- f(N) Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
Supplier: Ricca Chemical Company RV010932500N
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 97.52%,0.48% | |
| 1.01 | |
| NNaO2 | |
| Approximately 0°C | |
| [Na+].[O-]N=O | |
| 69.00 | |
| 23668193 | |
| High Purity |
| 7632-00-0 , 7732-18-5 | |
| 100%,0.50% | |
| Liquid | |
| MFCD00011118 | |
| LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
| sodium nitrite | |
| 500 mL | |
| CHEBI:78870 |
Chemical Identifiers
| 7632-00-0 | |
| 69.00 | |
| LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
| CHEBI:78870 | |
| [Na+].[O-]N=O |
| NNaO2 | |
| MFCD00011118 | |
| 23668193 | |
| sodium nitrite |