missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Nitrite(N) - NO2- f(N) Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Catalog No. p-7074057
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010932100 100 mL
RV010932250 250 mL
RV010932500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010932100 Supplier Ricca Chemical Company Supplier No. RV010932100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7632-00-0 , 7732-18-5
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.52%, 0.48%
CAS Max % 100%, 0.50%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity