missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Nitrite(N) - NO2- f(N) Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Facility

Supplier:  Ricca Chemical Company RV010932100N

 View more versions of this product

Catalog No. RV010932100


Only null left

Description

Description

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural LDPE Bottle
97.52%,0.48%
1.01
NNaO2
Approximately 0°C
[Na+].[O-]N=O
69.00
23668193
High Purity
7632-00-0 , 7732-18-5
100%,0.50%
Liquid
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
100 mL
CHEBI:78870

Chemical Identifiers

7632-00-0
69.00
LPXPTNMVRIOKMN-UHFFFAOYSA-M
CHEBI:78870
[Na+].[O-]N=O
NNaO2
MFCD00011118
23668193
sodium nitrite
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions