missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Nitrate(N) - NO3- f(N) Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Facility

Supplier:  Ricca Chemical Company RV010929500N

 View more versions of this product

Catalog No. RV010929500


Only null left

Description

Description

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
97.40%,0.60%
1.01
NNaO3
Approximately 0°C
[Na+].[O-][N+]([O-])=O
84.99
24268
High Purity
7631-99-4 , 7732-18-5
100%,0.62%
Liquid
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate
500 mL
CHEBI:63005

Chemical Identifiers

7631-99-4
84.99
VWDWKYIASSYTQR-UHFFFAOYSA-N
CHEBI:63005
[Na+].[O-][N+]([O-])=O
NNaO3
MFCD00011119
24268
sodium nitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions