missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Nickel Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical

Catalog No. RV010400500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010400500 500 mL
RV010400100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010400500 Supplier Ricca Chemical Company Supplier No. RV010400500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in 5% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 13138-45-9 , 7697-37-2 , 7732-18-5
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00011139
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
PubChem CID 25736
IUPAC Name nickel(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 90.05%, 4.90%, 3.05%
CAS Max % 93.73%, 5.10%, 3.17%
Specific Gravity 1
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity