missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Neodymium (Nd) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010036500N

 View more versions of this product

Catalog No. RV010036500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.75%,1.96%,0.29%
1.01
H12N3NdO15
VQVDTKCSDUNYBO-UHFFFAOYSA-N
neodymium(3+);trinitrate;hexahydrate
500 mL
High Purity
16454-60-7 , 7697-37-2 , 16454-60-7
99.65%,2.04%,0.31%
Liquid
Approximately 0°C
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
438.344
204494

Chemical Identifiers

16454-60-7
438.344
204494
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
H12N3NdO15
VQVDTKCSDUNYBO-UHFFFAOYSA-N
neodymium(3+);trinitrate;hexahydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions