missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Neodymium (Nd) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$356.90 - $726.37

Chemical Identifiers

CAS 16454-60-7
Molecular Formula H12N3NdO15
Molecular Weight (g/mol) 438.344
InChI Key VQVDTKCSDUNYBO-UHFFFAOYSA-N
PubChem CID 204494
IUPAC Name neodymium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010036100
View Documents
Ricca Chemical Company
RV010036100N
100 mL
Each for $356.90
Only null left
 
RV010036500
View Documents
Ricca Chemical Company
RV010036500N
500 mL
Each for $726.37
Only null left
 
Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

16454-60-7
438.344
204494
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
H12N3NdO15
VQVDTKCSDUNYBO-UHFFFAOYSA-N
neodymium(3+);trinitrate;hexahydrate

Specifications

Natural LDPE Bottle
95.75%,1.96%,0.29%
1.01
H12N3NdO15
VQVDTKCSDUNYBO-UHFFFAOYSA-N
neodymium(3+);trinitrate;hexahydrate
100 mL
High Purity
16454-60-7 , 7697-37-2 , 16454-60-7
99.65%,2.04%,0.31%
Liquid
Approximately 0°C
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
438.344
204494
Videos
SDS
Documents

Documents