missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Molybdenum (Mo) Standard for AAS 1000 ppm in H2O, Ricca Chemical

Catalog No. RV010079100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010079100 100 mL
RV010079500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010079100 Supplier Ricca Chemical Company Supplier No. RV010079100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 12027-67-7 , 7732-18-5
Molecular Formula H24Mo7N6O24
Molecular Weight (g/mol) 1163.85
MDL Number MFCD00064633
InChI Key MISTUIVWPMGXFX-UHFFFAOYSA-T
PubChem CID 71306766
IUPAC Name hexaammonium dodecaoxododecacyclo[7.5.1.13,5.13,13.15,7.17,9.01,4.01,6.01,8.02,13.011,14.011,15]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium)
SMILES [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.83%, 0.17%
CAS Max % 100%, 0.17%
Specific Gravity 0.999
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity