accessibility menu, dialog, popup

UserName

CAS RN 12027-67-7

MoOOOOOOMoMoOOOOOOOMoMoOOOOOOMoMoOOOOONH4NH4NH4NH4NH4NH4

CAS RN 12027-67-7

IUPAC Name: hexaammonium dodecaoxododecacyclo[7.5.1.1³,⁵.1³,¹³.1⁵,⁷.1⁷,⁹.0¹,⁴.0¹,⁶.0¹,⁸.0²,¹³.0¹¹,¹⁴.0¹¹,¹⁵]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium)
Synonyms: hexaammonium dodecaoxododecacyclo[7.5.1.1³,⁵.1³,¹³.1⁵,⁷.1⁷,⁹.0¹,⁴.0¹,⁶.0¹,⁸.0²,¹³.0¹¹,¹⁴.0¹¹,¹⁵]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium)
Molecular Weight (g/mol): 1163.85
Molecular Formula: H24Mo7N6O24
InChi Key: MISTUIVWPMGXFX-UHFFFAOYSA-T
SMILES: [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11
Grade 
  • (11)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results