missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Mercury Standard, 1.557 mg/Kg in H2O, Ricca Chemical

Catalog No. RV010762100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010762100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010762100 Supplier Ricca Chemical Company Supplier No. RV010762100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10045-94-0 , 7697-37-2 , 7732-18-5
Molecular Formula HgN2O6
Molecular Weight (g/mol) 324.60
MDL Number MFCD00011038
InChI Key ORMNPSYMZOGSSV-UHFFFAOYSA-N
PubChem CID 24864
IUPAC Name mercury(2+) dinitrate
SMILES [Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 1.96%, 0.00%
CAS Max % 2.04%, 0.00%
Specific Gravity 1.06
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade High Purity
Chemical Name or Material Mercury Standard
Show More Show Less