missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Manganese Standard for ICP 10 g/L in 5% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010395100N

Catalog No. RV010395100

Only null left
Explore available promotions

Description

Description

  • 10 g/L in 5% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Manganese ICP Standard
10377-66-9 , 7647-01-0 , 7732-18-5
94.56%,5.10%
Liquid
UN3264
<2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
MIVBAHRSNUNMPP-UHFFFAOYSA-N
manganese(2+);dinitrate
100 mL
High Purity
Natural LDPE Bottle
90.86%,4.90%
1
MnN2O6
Colorless
Approximately 0°C
Miscible with water
Standard
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2]
178.946
61511
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions