missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Manganese Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$461.10 - $1069.67
Chemical Identifiers
| CAS | 10377-66-9 |
|---|---|
| Molecular Formula | MnN2O6 |
| Molecular Weight (g/mol) | 178.946 |
| InChI Key | MIVBAHRSNUNMPP-UHFFFAOYSA-N |
| PubChem CID | 61511 |
| IUPAC Name | manganese(2+);dinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010293100
|
Ricca Chemical Company
RV010293100N |
100 mL |
Each for $461.10
|
|
|||||
|
RV010293250
|
Ricca Chemical Company
RV010293250N |
250 mL |
Each for $751.53
|
|
|||||
|
RV010293500
|
Ricca Chemical Company
RV010293500N |
500 mL |
Each for $1,069.67
|
|
|||||
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10377-66-9 | |
| 178.946 | |
| 61511 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2] |
| MnN2O6 | |
| MIVBAHRSNUNMPP-UHFFFAOYSA-N | |
| manganese(2+);dinitrate |
Specifications
| Natural LDPE Bottle | |
| 95.81%,1.96% | |
| 1 | |
| MnN2O6 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2] | |
| 178.946 | |
| 61511 |
| 10377-66-9 , 7647-01-0 , 7732-18-5 | |
| 99.73%,2.04% | |
| Liquid | |
| Approximately 0°C | |
| MIVBAHRSNUNMPP-UHFFFAOYSA-N | |
| manganese(2+);dinitrate | |
| 100 mL | |
| High Purity |