missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Manganese (Mn) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$246.03 - $433.43
Chemical Identifiers
| CAS | 10377-66-9 |
|---|---|
| Molecular Formula | MnN2O6 |
| Molecular Weight (g/mol) | 178.946 |
| InChI Key | MIVBAHRSNUNMPP-UHFFFAOYSA-N |
| PubChem CID | 61511 |
| IUPAC Name | manganese(2+);dinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010078100
|
Ricca Chemical Company
RV010078100N |
100 mL |
Each for $246.03
|
|
|||||
|
RV010078500
|
Ricca Chemical Company
RV010078500N |
500 mL |
Each for $433.43
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10377-66-9 | |
| 178.946 | |
| 61511 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2] |
| MnN2O6 | |
| MIVBAHRSNUNMPP-UHFFFAOYSA-N | |
| manganese(2+);dinitrate |
Specifications
| Natural LDPE Bottle | |
| 95.72%,1.96%,0.32% | |
| 1.01 | |
| MnN2O6 | |
| MIVBAHRSNUNMPP-UHFFFAOYSA-N | |
| manganese(2+);dinitrate | |
| 100 mL | |
| High Purity |
| 10377-66-9 , 7697-37-2 , 7732-18-5 | |
| 99.62%,2.04%,0.34% | |
| Liquid | |
| Approximately 0°C | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2] | |
| 178.946 | |
| 61511 |