missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Manganese (Mn) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010032500N

 View more versions of this product

Catalog No. RV010032500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.81%,1.96%
1.01
MnN2O6
MIVBAHRSNUNMPP-UHFFFAOYSA-N
manganese(2+);dinitrate
500 mL
High Purity
10377-66-9 , 7647-01-0 , 7732-18-5
99.73%,2.04%
Liquid
Approximately 0°C
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2]
178.946
61511

Chemical Identifiers

10377-66-9
178.946
61511
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2]
MnN2O6
MIVBAHRSNUNMPP-UHFFFAOYSA-N
manganese(2+);dinitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions