missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Lithium (Li) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$249.47 - $439.50
Chemical Identifiers
| CAS | 7790-69-4 |
|---|---|
| Molecular Formula | LiNO3 |
| Molecular Weight (g/mol) | 68.94 |
| MDL Number | MFCD00011094 |
| InChI Key | IIPYXGDZVMZOAP-UHFFFAOYSA-N |
| PubChem CID | 10129889 |
| ChEBI | CHEBI:63315 |
| IUPAC Name | lithium(1+) nitrate |
| SMILES | [Li+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010030100
|
Ricca Chemical Company
RV010030100N |
100 mL |
Each for $249.47
|
|
|||||
|
RV010030500
|
Ricca Chemical Company
RV010030500N |
500 mL |
Each for $439.50
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7790-69-4 | |
| 68.94 | |
| IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
| CHEBI:63315 | |
| [Li+].[O-][N+]([O-])=O |
| LiNO3 | |
| MFCD00011094 | |
| 10129889 | |
| lithium(1+) nitrate |
Specifications
| Natural LDPE Bottle | |
| 95.07%,1.96%,0.97% | |
| 1.01 | |
| LiNO3 | |
| Approximately 0°C | |
| [Li+].[O-][N+]([O-])=O | |
| 68.94 | |
| 10129889 | |
| High Purity |