missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iron Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$429.56 - $429.56
Chemical Identifiers
| CAS | 10421-48-4 |
|---|---|
| Molecular Formula | FeN3O9 |
| Molecular Weight (g/mol) | 241.857 |
| InChI Key | VCJMYUPGQJHHFU-UHFFFAOYSA-N |
| PubChem CID | 25251 |
| IUPAC Name | iron(3+);trinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010588100
|
Ricca Chemical Company
RV010588100N |
100 mL |
Each for $429.56
|
|
|||||
Description
- 10 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10421-48-4 | |
| 241.857 | |
| 25251 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3] |
| FeN3O9 | |
| VCJMYUPGQJHHFU-UHFFFAOYSA-N | |
| iron(3+);trinitrate |
Specifications
| Natural LDPE Bottle | |
| 96.04%,1.96%,0.00% | |
| 1 | |
| FeN3O9 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3] | |
| 241.857 | |
| 25251 |
| 10421-48-4 , 7697-37-2 , 7732-18-5 | |
| 99.96%,2.04%,0.00% | |
| Liquid | |
| Approximately 0°C | |
| VCJMYUPGQJHHFU-UHFFFAOYSA-N | |
| iron(3+);trinitrate | |
| 100 mL | |
| High Purity |