missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iron Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010451250N
Description
- 10 g/L in 5% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 88.86%,4.90%,4.24% | |
| 1 | |
| FeN3O9 | |
| Approximately 0°C | |
| VCJMYUPGQJHHFU-UHFFFAOYSA-N | |
| iron(3+);trinitrate | |
| 250 mL | |
| High Purity |
| 10421-48-4 , 7697-37-2 , 7732-18-5 | |
| 92.48%,5.10%,4.42% | |
| Liquid | |
| UN3264 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3] | |
| 241.857 | |
| 25251 |
Chemical Identifiers
| 10421-48-4 | |
| 241.857 | |
| 25251 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3] |
| FeN3O9 | |
| VCJMYUPGQJHHFU-UHFFFAOYSA-N | |
| iron(3+);trinitrate |