missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iron Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical
Description
- 10 g/L in 5% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 88.86%, 4.90%, 4.24% |
| CAS Max % | 92.48%, 5.10%, 4.42% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| UN Number | UN3264 |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 100 mL |
| Grade | High Purity |