missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iron (Fe) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 95.62%, 1.96%, 0.42% |
| CAS Max % | 99.52%, 2.04%, 0.44% |
| Specific Gravity | 1.016 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Type | Single-element Standard Solution |
| Quantity | 500 mL |
| Grade | High Purity |
| Chemical Name or Material | Iron(III) Nitrate |