missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iridium Standard for ICP 1000 ppm in 10% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010287500N
Description
- 1000 ppm in 10% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 88.04%,9.80%,0.16% | |
| 1 | |
| Cl3Ir | |
| Approximately 0°C | |
| DANYXEHCMQHDNX-UHFFFAOYSA-K | |
| trichloroiridium | |
| 500 mL | |
| High Purity |
| 10025-83-9,7647-01-0,7732-18-5 | |
| 91.64%,10.20%,0.16% | |
| Liquid | |
| UN1789 | |
| Standard | |
| Cl[Ir](Cl)Cl | |
| 298.567 | |
| 25563 |
Chemical Identifiers
| 10025-83-9 | |
| 298.567 | |
| 25563 | |
| Cl[Ir](Cl)Cl |
| Cl3Ir | |
| DANYXEHCMQHDNX-UHFFFAOYSA-K | |
| trichloroiridium |