missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Holmium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010285100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010285100 100 mL
RV010285250 250 mL
RV010285500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010285100 Supplier Ricca Chemical Company Supplier No. RV010285100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 14483-18-2 , 7697-37-2 , 7732-18-5
Molecular Formula H10HoN3O14
Molecular Weight (g/mol) 441.017
InChI Key BJUGWWDCFYEYOA-UHFFFAOYSA-N
PubChem CID 25021754
IUPAC Name holmium(3+);trinitrate;pentahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.78%, 1.96%, 0.26%
CAS Max % 99.68%, 2.04%, 0.28%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity