missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Catalog No. RV010004500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010004500 500 mL
RV010004100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010004500 Supplier Ricca Chemical Company Supplier No. RV010004500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 16903-35-8 , 7647-01-0 , 7732-18-5

Specifications

Packaging Amber Glass Bottle
CAS Min % 95.87%, 1.96%, 0.17%
CAS Max % 99.79%, 2.04%, 0.17%
Specific Gravity 1.024
Physical Form Liquid
Melting Point Approximately 0°C
Type Single-element Standard Solution
Quantity 500 mL
Grade High Purity
Chemical Name or Material Gold