missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Gallium (Ga) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010020500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010020500 500 mL
RV010020100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010020500 Supplier Ricca Chemical Company Supplier No. RV010020500N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 69365-72-6 , 7697-37-2 , 7732-18-5
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
PubChem CID 11550823
IUPAC Name gallium(3+) trinitrate
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 95.68%, 1.96%, 0.36%
CAS Max % 99.58%, 2.04%, 0.38%
Specific Gravity 1.009
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 500 mL
Grade High Purity