missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Gallium (Ga) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010020100
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010020100 100 mL
RV010020500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. RV010020100 missing translation for 'mfr' Ricca Chemical Company missing translation for 'supplierNo' RV010020100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 69365-72-6 , 7697-37-2 , 7732-18-5
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
PubChem CID 11550823
IUPAC Name gallium(3+) trinitrate
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.68%, 1.96%, 0.36%
CAS Max % 99.58%, 2.04%, 0.38%
Specific Gravity 1.009
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity