missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Erbium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$410.91 - $1054.34
Chemical Identifiers
| CAS | 10031-51-3 |
|---|---|
| Molecular Formula | ErH10N3O14 |
| Molecular Weight (g/mol) | 443.35 |
| MDL Number | MFCD00149691 |
| InChI Key | LWHHUEHWVBVASY-UHFFFAOYSA-N |
| PubChem CID | 53249207 |
| IUPAC Name | erbium(3+) pentahydrate trinitrate |
| SMILES | O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010277100
|
Ricca Chemical Company
RV010277100N |
100 mL |
Each for $410.91
|
|
|||||
|
RV010277250
|
Ricca Chemical Company
RV010277250N |
250 mL |
Each for $737.50
|
|
|||||
|
RV010277500
|
Ricca Chemical Company
RV010277500N |
500 mL |
Each for $1,054.34
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10031-51-3 | |
| 443.35 | |
| LWHHUEHWVBVASY-UHFFFAOYSA-N | |
| erbium(3+) pentahydrate trinitrate |
| ErH10N3O14 | |
| MFCD00149691 | |
| 53249207 | |
| O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Natural LDPE Bottle | |
| 95.78%,1.96%,0.26% | |
| 1 | |
| ErH10N3O14 | |
| Approximately 0°C | |
| LWHHUEHWVBVASY-UHFFFAOYSA-N | |
| erbium(3+) pentahydrate trinitrate | |
| 100 mL | |
| High Purity |
| 10031-51-3,7697-37-2,7732-18-5 | |
| 99.68%,2.04%,0.28% | |
| Liquid | |
| MFCD00149691 | |
| Standard | |
| O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 443.35 | |
| 53249207 |