missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Copper (Cu) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010015500N

 View more versions of this product

Catalog No. RV010015500


Only null left

Description

Description

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.83%,1.96%,0.21%
1.01
Approximately 0°C
High Purity
7447-39-4 , 7647-01-0 , 7732-18-5
99.75%,2.04%,0.21%
Liquid
500 mL

Chemical Identifiers

7447-39-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions