missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Cobalt Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$467.57 - $1084.63
Chemical Identifiers
| CAS | 10141-05-6 |
|---|---|
| Molecular Formula | CoN2O6 |
| Molecular Weight (g/mol) | 182.941 |
| InChI Key | UFMZWBIQTDUYBN-UHFFFAOYSA-N |
| PubChem CID | 25000 |
| ChEBI | CHEBI:86209 |
| IUPAC Name | cobalt(2+);dinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010272100
|
Ricca Chemical Company
RV010272100N |
100 mL |
Each for $467.57
|
|
|||||
|
RV010272250
|
Ricca Chemical Company
RV010272250N |
250 mL |
Each for $762.07
|
|
|||||
|
RV010272500
|
Ricca Chemical Company
RV010272500N |
500 mL |
Each for $1,084.63
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10141-05-6 | |
| 182.941 | |
| 25000 | |
| cobalt(2+);dinitrate |
| CoN2O6 | |
| UFMZWBIQTDUYBN-UHFFFAOYSA-N | |
| CHEBI:86209 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2] |
Specifications
| Natural LDPE Bottle | |
| 95.74%,1.96%,0.30% | |
| 1 | |
| CoN2O6 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2] | |
| 182.941 | |
| 25000 | |
| High Purity |
| 10141-05-6 , 7697-37-2 , 7732-18-5 | |
| 99.64%,2.04%,0.32% | |
| Liquid | |
| Approximately 0°C | |
| UFMZWBIQTDUYBN-UHFFFAOYSA-N | |
| cobalt(2+);dinitrate | |
| 100 mL | |
| CHEBI:86209 |