missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cobalt (Co) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010012500N

 View more versions of this product

Catalog No. RV010012500

Only null left

Description

Description

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.82%,1.96%,0.22%
1.01
Cl2Co
Approximately 0°C
[Cl-].[Cl-].[Co++]
129.83
High Purity
7646-79-9 , 7647-01-0 , 7732-18-5
99.74%,2.04%,0.22%
Liquid
MFCD00010938
GVPFVAHMJGGAJG-UHFFFAOYSA-L
λ2-cobalt(2+) dichloride
500 mL

Chemical Identifiers

7646-79-9
129.83
GVPFVAHMJGGAJG-UHFFFAOYSA-L
[Cl-].[Cl-].[Co++]
Cl2Co
MFCD00010938
λ2-cobalt(2+) dichloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions