missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Chromium (Cr) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 95.75%, 1.96%, 0.30% |
| CAS Max % | 99.65%, 2.04%, 0.32% |
| Specific Gravity | 1.01 |
| Physical Form | Liquid |
| Molecular Formula | Cl3CrH12O6 |
| Melting Point | Approximately 0°C |
| Type | Single-element Standard Solution |
| InChI Key | LJAOOBNHPFKCDR-UHFFFAOYSA-K |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
| Show More |