missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cesium - Cs+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Facility

Supplier:  Ricca Chemical Company RV010908500N

 View more versions of this product

Catalog No. RV010908500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
97.85%,0.15%
1.01
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
500 mL
High Purity
7789-18-6 , 7732-18-5
100%,0.15%
Liquid
Approximately 0°C
[N+](=O)([O-])[O-].[Cs+]
194.909
62674

Chemical Identifiers

7789-18-6
194.909
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions