missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010014500N
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 95.89%,1.96%,0.15% | |
| 1.01 | |
| CsNO3 | |
| NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
| cesium;nitrate | |
| 500 mL | |
| High Purity |
| 7789-18-6,7697-37-2,7732-18-5 | |
| 99.81%,2.04%,0.15% | |
| Liquid | |
| Approximately 0°C | |
| [N+](=O)([O-])[O-].[Cs+] | |
| 194.909 | |
| 62674 |
Chemical Identifiers
| 7789-18-6 | |
| 194.909 | |
| 62674 | |
| [N+](=O)([O-])[O-].[Cs+] |
| CsNO3 | |
| NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
| cesium;nitrate |