missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. p-7239347
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RV010014100 100 mL Natural LDPE Bottle
RV010014500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010014100 Supplier Ricca Chemical Company Supplier No. RV010014100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7789-18-6 , 7697-37-2 , 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.85, 2, 0.15
CAS Max % 97.85, 2, 0.15
Density 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material VeriSpec™ Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO3, Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
ShelfLife 24 months
Recommended Storage Ambient