missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010014100N

 View more versions of this product

Catalog No. RV010014100


Only null left
Explore available promotions

Description

Description

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

VeriSpec™ Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO3
Natural LDPE Bottle
97.85,2,0.15
1.01
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
100 mL
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
7789-18-6,7697-37-2,7789-18-6
97.85,2,0.15
Liquid
Approximately 0°C
[N+](=O)([O-])[O-].[Cs+]
194.909
62674

Chemical Identifiers

7789-18-6
194.909
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions