missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Calcium - Ca2+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
$326.47 - $600.84
Chemical Identifiers
| CAS | 10124-37-5 |
|---|---|
| Molecular Formula | CaN2O6 |
| Molecular Weight (g/mol) | 164.086 |
| InChI Key | ZCCIPPOKBCJFDN-UHFFFAOYSA-N |
| PubChem CID | 24963 |
| ChEBI | CHEBI:64205 |
| IUPAC Name | calcium;dinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010907100
|
Ricca Chemical Company
RV010907100N |
100 mL |
Each for $326.47
|
|
|||||
|
RV010907250
|
Ricca Chemical Company
RV010907250N |
250 mL |
Each for $421.72
|
|
|||||
|
RV010907500
|
Ricca Chemical Company
RV010907500N |
500 mL |
Each for $600.84
|
|
|||||
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10124-37-5 | |
| 164.086 | |
| 24963 | |
| calcium;dinitrate |
| CaN2O6 | |
| ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
| CHEBI:64205 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2] |
Specifications
| Natural LDPE Bottle | |
| 97.60%,0.40% | |
| 1.01 | |
| CaN2O6 | |
| ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
| calcium;dinitrate | |
| 100 mL | |
| CHEBI:64205 |
| 10124-37-5 , 7732-18-5 | |
| 100%,0.42% | |
| Liquid | |
| Approximately 0°C | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2] | |
| 164.086 | |
| 24963 | |
| High Purity |