missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Cadmium (Cd) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$220.66 - $388.69
Chemical Identifiers
| CAS | 10325-94-7 |
|---|---|
| Molecular Formula | CdN2O6 |
| Molecular Weight (g/mol) | 236.422 |
| InChI Key | XIEPJMXMMWZAAV-UHFFFAOYSA-N |
| PubChem CID | 25154 |
| ChEBI | CHEBI:77732 |
| IUPAC Name | cadmium(2+);dinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cd+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010069100
|
Ricca Chemical Company
RV010069100N |
100 mL |
Each for $220.66
|
|
|||||
|
RV010069500
|
Ricca Chemical Company
RV010069500N |
500 mL |
Each for $388.69
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10325-94-7 | |
| 236.422 | |
| 25154 | |
| cadmium(2+);dinitrate |
| CdN2O6 | |
| XIEPJMXMMWZAAV-UHFFFAOYSA-N | |
| CHEBI:77732 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cd+2] |
Specifications
| Natural LDPE Bottle | |
| 95.83%,1.96%,0.21% | |
| 1.01 | |
| CdN2O6 | |
| XIEPJMXMMWZAAV-UHFFFAOYSA-N | |
| cadmium(2+);dinitrate | |
| 100 mL | |
| CHEBI:77732 |
| 10325-94-7,7697-37-2,7732-18-5 | |
| 99.75%,2.04%,0.21% | |
| Liquid | |
| Approximately 0°C | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cd+2] | |
| 236.422 | |
| 25154 | |
| High Purity |