missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Barium Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010266250N
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 95.89%,1.96%,0.15% | |
| 1 | |
| BaCl2 | |
| Approximately 0°C | |
| WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
| barium(2+) dichloride | |
| 250 mL | |
| CHEBI:63317 |
| 10361-37-2 , 7647-01-0 , 7732-18-5 | |
| 99.81%,2.04%,0.15% | |
| Liquid | |
| MFCD00003445 | |
| Standard | |
| [Cl-].[Cl-].[Ba++] | |
| 208.23 | |
| 25204 | |
| High Purity |
Chemical Identifiers
| 10361-37-2 | |
| 208.23 | |
| WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
| CHEBI:63317 | |
| [Cl-].[Cl-].[Ba++] |
| BaCl2 | |
| MFCD00003445 | |
| 25204 | |
| barium(2+) dichloride |