missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ As3+ Standard for ICP 1000 ppm, Ricca Chemical

Catalog No. RV010643100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010643100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010643100 Supplier Ricca Chemical Company Supplier No. RV010643100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS 7784-46-5, 1310-73-2, 7647-14-5, 7732-18-5
CAS Min % 97.25%, 0.49%, 0.17%, 0.10%
CAS Max % 100%, 0.51%, 0.17%, 0.10%
Specific Gravity 1
Physical Form Liquid
Molecular Formula AsNaO2
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES [O-][As]=O.[Na+]
IUPAC Name sodium;oxoarsinite
Molecular Weight (g/mol) 129.909
Quantity 100 mL
PubChem CID 443495
ChEBI CHEBI:29678
Grade High Purity
Chemical Name or Material Arsenic ICP Standard
Show More Show Less