missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Arsenic Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 96.02%, 1.96%, 0.02% |
| CAS Max % | 99.94%, 2.04%, 0.02% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | Approximately 0°C |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solubility | Miscible with water |
| Solution Type | Calibration Standard |
| Show More |