missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Arsenic (As) Standard for AAS 20 ppb in 1% HNO3, Ricca Chemical

Catalog No. RV01012250N
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV01012250N 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV01012250N Supplier Ricca Chemical Company Supplier No. RV01012250N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7778-39-4 , 7697-37-2 , 7732-18-5
Molecular Formula AsH3O4
Molecular Weight (g/mol) 141.942
InChI Key DJHGAFSJWGLOIV-UHFFFAOYSA-N
PubChem CID 234
ChEBI CHEBI:18231
IUPAC Name arsoric acid
SMILES O[As](=O)(O)O

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.02%, 0.98%
CAS Max % 100%, 1.02%
Specific Gravity 1.01
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-element Standard Solution
Quantity 50 mL
Grade High Purity
Chemical Name or Material Arsenic AAS Standard
Show More Show Less