missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Arsenic (As) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$246.03 - $433.43
Chemical Identifiers
| CAS | 7778-39-4 |
|---|---|
| Molecular Formula | AsH3O4 |
| Molecular Weight (g/mol) | 141.942 |
| InChI Key | DJHGAFSJWGLOIV-UHFFFAOYSA-N |
| PubChem CID | 234 |
| ChEBI | CHEBI:18231 |
| IUPAC Name | arsoric acid |
| SMILES | O[As](=O)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010065100
|
Ricca Chemical Company
RV010065100N |
100 mL |
Each for $246.03
|
|
|||||
|
RV010065500
|
Ricca Chemical Company
RV010065500N |
500 mL |
Each for $433.43
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7778-39-4 | |
| 141.942 | |
| 234 | |
| arsoric acid |
| AsH3O4 | |
| DJHGAFSJWGLOIV-UHFFFAOYSA-N | |
| CHEBI:18231 | |
| O[As](=O)(O)O |
Specifications
| Natural LDPE Bottle | |
| 95.85%,1.96%,0.19% | |
| 1.015 | |
| AsH3O4 | |
| DJHGAFSJWGLOIV-UHFFFAOYSA-N | |
| arsoric acid | |
| 100 mL | |
| CHEBI:18231 |