missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Antimony (Sb) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical

Catalog No. p-7073829
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010047100 100 mL
RV010047500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010047100 Supplier Ricca Chemical Company Supplier No. RV010047100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 20% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10025-91-9 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name antimony(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 78.21%, 19.60%, 0.19%
CAS Max % 81.41%, 20.40%, 0.19%
Specific Gravity 1.02
Physical Form Liquid
UN Number UN1789
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity