missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Ammonium Nitrate Matrix Modifier Standard for AAS 50,000 ppm in H2O, Ricca Chemical

Catalog No. RV01013250N
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV01013250N 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV01013250N Supplier Ricca Chemical Company Supplier No. RV01013250N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 6484-52-2 , 7732-18-5
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.10%, 4.90%
CAS Max % 96.90%, 5.10%
Specific Gravity 1.01
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Matrix Modifier
Quantity 50 mL
Grade High Purity
Chemical Name or Material Ammonium Nitrate AAS Matrix Modifier
Show More Show Less