missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Ammonium - NH4+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
$331.03 - $598.78
Chemical Identifiers
| CAS | 6484-52-2 |
|---|---|
| Molecular Formula | H4N2O3 |
| Molecular Weight (g/mol) | 80.04 |
| MDL Number | MFCD00011425 |
| InChI Key | DVARTQFDIMZBAA-UHFFFAOYSA-O |
| PubChem CID | 22985 |
| ChEBI | CHEBI:63038 |
| IUPAC Name | ammonium nitrate |
| SMILES | [NH4+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010901100
|
Ricca Chemical Company
RV010901100N |
100 mL |
N/A
|
|
|||||
|
RV010901250
|
Ricca Chemical Company
RV010901250N |
250 mL |
N/A
|
|
|||||
|
RV010901500
|
Ricca Chemical Company
RV010901500N |
500 mL |
N/A
|
|
|||||
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 6484-52-2 | |
| 80.04 | |
| DVARTQFDIMZBAA-UHFFFAOYSA-O | |
| CHEBI:63038 | |
| [NH4+].[O-][N+]([O-])=O |
| H4N2O3 | |
| MFCD00011425 | |
| 22985 | |
| ammonium nitrate |
Specifications
| Natural LDPE Bottle | |
| 97.57%,0.43% | |
| 1.01 | |
| H4N2O3 | |
| Approximately 0°C | |
| [NH4+].[O-][N+]([O-])=O | |
| 80.04 | |
| 22985 | |
| High Purity |